Starter pH Pen Meters
Starter pH Pen Meters
Actual product may vary
Starter pH Pen Meters
Convenient pH testing. OHAUS Starter pH Pen Meters offer a convenient way to measure pH, or the relative acidity or alkalinity of a sample at a given temperature. The pH pen meters can be completely operated with just one hand, freeing up your other hand to handle samples.
Have Questions?
Catalog NumberAccuracy (pH)Range (pH)Resolution (pH)Display Type
OHA30073971±0.10 to 140.01Single Line
Catalog number OHA30073971
Price (MYR)
953.00
Add to cart
Accuracy (pH):
±0.1
Range (pH):
0 to 14
Resolution (pH):
0.01
Display Type:
Single Line
Price (MYR)
953.00
Add to cart
  • Fast, straightforward results
  • Durable IP67 waterproof design
  • Equipped with wrist strap for security
  • Automatic Shutdown after six minutes of non-use
  • Requires four AG13 1.5V Micro Alkaline Batteries
  • Size/Weight: 185 x 45 x 38 mm, 110 g
  • Includes buffer powder pack (ST20)
Specifications
Accuracy (pH)±0.1
Includes4 1.5V Batteries
For Use With (Application)Measuring pH, or the relative acidity or alkalinity of a sample at a given temperature
Measurement ModespH with LogR Temperature
CalibrationAutomatic/Manual
Range (pH)0 to 14
Resolution (pH)0.01
Calibration Points3
Display TypeSingle Line
Backlight OptionNone
Data Points30
Log Function TypeAutomatic Data Logging
Weight (Metric)110 g
Weight (English)0.24 lb.
Battery Type4 x AG13 1.5 V Micro Alkaline
TypePortable pH Meter
Unit Size