BioClean™ Nitrile Cleanroom RABS/Isolator Gloves
BioClean™ Nitrile Cleanroom RABS/Isolator Gloves
Actual product may vary
BioClean™ Nitrile Cleanroom RABS/Isolator Gloves
BioClean™ Nitrile Clean RABS/Isolator Gloves are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties. Designed for use in injectable manufacturing and pharmaceutical production areas, our CGL series of gloves are cleanroom processed and packed.
Have Questions?
Catalog NumberPort Size (English)Port Size (Metric)
NRTCGL36NIT5912 to 14 in.30.5 to 35.6 cm
Catalog number NRTCGL36NIT59
Price (MYR)
34,693.00
CS
Add to cart
Port Size (English):
12 to 14 in.
Port Size (Metric):
30.5 to 35.6 cm
Price (MYR)
34,693.00
CS
Add to cart
  • Tested against ASTM D6978-05 for handling chemo drugs.
  • 100% water leak tested.
  • Ultra-clean surface ensures product protection.
  • Specially designed to minimize hand fatigue.
  • Autoclave compatible.
  • ISO 4 Cleanroom laundered and packaged.
  • Resistant to VHP, IPA, and disinfectants.
Specifications
Certifications/ComplianceISO 4
ColorWhite
Laboratory/Cleanroom UseCleanroom
Latex FreeYes
MaterialNitrile
Size9.75
Thickness (English)24 mil
Thickness (Metric)0.60 mm
ModelBioClean™ Clean Nitrile RABS / Insulator Gloves CGL
Port Size (English)12 to 14 in.
Port Size (Metric)30.5 to 35.6 cm
SeriesBioClean™ Clean Nitrile RABS / Insulator Gloves CGL
Length (English)33 in.
Length (Metric)84 cm
Unit SizeCS