Economy Muffle Furnace 1100 °C With LE 1/11, Flap Door and Controller R7
Economy Muffle Furnace 1100 °C With LE 1/11, Flap Door and Controller R7
Actual product may vary
Economy Muffle Furnace 1100 °C With LE 1/11, Flap Door and Controller R7
With their convincing price/performance ratio and the fast heat-up rates, these compact muffle furnaces are perfect for many applications in the laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Dimensions (D x W x H) InteriorDimensions (L x W x H) Exterior
NABLE062K17N16 L200 x 170 x 170 mm435 x 390 x 490 mm
NABLE241K17N124 L330 x 260 x 285 mm570 x 490 x 585 mm
Catalog number NABLE062K17N1
Price (MYR)
17,497.00
Add to cart
Capacity (Metric):
6 L
Dimensions (D x W x H) Interior:
200 x 170 x 170 mm
Dimensions (L x W x H) Exterior:
435 x 390 x 490 mm
Price (MYR)
17,497.00
Add to cart
  • Only use fiber materials not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Heating from two sides from heating elements protected in quartz glass tubes
  • Maintenance-friendly replacement of heating elements and insulation
  • Adjustable air inlet integrated in door
  • Exhaust air outlet in rear wall of furnace
  • Compact dimensions and light weight
  • Short delivery times for standard furnaces and spare parts
Additional equipment:
  • Chimney, chimney with fan or catalytic converter
Specifications
Capacity (Metric)6 L
Temperature (Metric)1100°C
TypeFurnace
StyleFlap door
Temperature ControlR7
Dimensions (D x W x H) Interior200 x 170 x 170 mm
Dimensions (L x W x H) Exterior435 x 390 x 490 mm
Weight (Metric)18 kg
Electrical Requirements1-phase
Unit Size