Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader
Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader
Actual product may vary
Thermo Fisher Scientific
Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader
Various accessories for use with the Varioskan™ LUX Multimode Microplate Reader.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
N0269396-well adapter for plate with lid
VLUPD005Dispenser Upgrade Kit
VLUPT004TRF Upgrade Kit
N026976-48-well adapter for plate with lid
SP-00094Dispensing tube assembly for a dispenser
Catalog number N02693
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
96-well adapter for plate with lid
Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.
Varioskan LUX is a modular instrument, you can upgrade it with missing features later.
  • Installation of the upgrade kits must be carried out by trained and authorized service personnel.
  • For TRF and/or AlphaScreen technology upgrades, the instrument must be equipped with a luminescence module.
  • TRF and/or AlphaScreen technology upgrades include excitation filters but not emission filters. Please order emission filters separately.
  • Please contact your local representative to discuss about the instrument upgrade option
Plate adapters are used for adapting plates that are of different heights.
  • Adapters adjust the plates to the optimum height for measuring and dispensing.
  • Choose the adapter based on the plate format in use and whether the lid is needed or not.
There are two types of plate trays: universal and robotic.
  • The universal plate tray is for basic use and it is compatible with all plate formats (6- to 1536-well plates).
  • A robotic plate tray is for automated use with robots and is compatible with 96- to 1536-well plate formats.
If the dispensers are in frequent use, it is recommended to replace the aspirate and dispensing tubings once a year.
Specifications
Product Type96-well adapter for plate with lid
For Use With (Equipment)Varioskan LUX Microplate Reader
Plate Type96-well Plate
Product LineVarioskan™
Unit Size