ACME Thread Plastic Handle
ACME Thread Plastic Handle
Actual product may vary
ACME Thread Plastic Handle
Plastic coated fiberglass, fixed length and extendable handles. Designed with ACME Connect threaded ends for use with the entire range of Micronova adapters, mop heads and tools for simplified and versatile cleaning.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Product Type
MICSTAEH-036WH46 to 91 cmExtendable Handle
MICSTAEH-183WH172.7 to 464.8 cmExtendable Handle
MICSTAH-60WH152.4 cmFixed Handle
Catalog number MICSTAEH-036WH
Price (MYR)
813.00
Add to cart
Length (Metric):
46 to 91 cm
Product Type:
Extendable Handle
Price (MYR)
813.00
Add to cart
Lightweight, plastic-coated fiberglass handles
Micronova Plastic ACME Threaded Handles are made from lightweight fiberglass coated in plastic and are available in a range of extendable and fixed length options. The ACME Connect threaded ends attach directly to the plastic (QP series) adapters, or use the SSA-2 ACME Thread Adapters to attach to Quick Disconnect (QD series) mop heads.
  • Lightweight, plastic coated fiberglass handles
  • ACME Connect threaded end connection
  • Available fixed length or extendable
  • Fixed lengths range from 12 to 60 inches (30 cm to 1.5 m)
  • Extendable maximum lengths range from 36 in to 11.5 ft (90 cm to 3.5 m)
  • ACME Connect threaded end attaches directly to the plastic (QP series) adapters
  • Attach to (QD series) mop heads via the SSA-2 ACME Thread Adapters
  • Autoclavable
Specifications
Color CodeWhite
Product TypeExtendable Handle
ColorWhite
Length (English)18 to 36 in.
SterilityNon-sterile
Length (Metric)46 to 91 cm
AutoclavableAutoclavable
Width (Metric)1.9 cm to 2.5 cm
Width (English)0.75 in. to 1 in.
Resistant ToCommon disinfectants and cleaning solutions
DisposableReusable
Thread StyleACME
Quantity1 Ea.
Care InstructionsStore at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
For Use With (Equipment)QP mop adapters and brushes with threaded connection
For Use With (Application)Mop Handle for Cleaning and Sanitizing Cleanrooms
Certifications/ComplianceISO 9001 Certified Facility Certificate of Conformance
Color-codedYes
MaterialFiberglass, Plastic
Diameter (Metric)1.9 cm to 2.5 cm
Unit Size