Irradiated Cleanroom Pen
Irradiated Cleanroom Pen
Actual product may vary
Irradiated Cleanroom Pen
Low-sodium, indelible ink ballpoint pens feature a unique polypropylene barrel that limits particle generation and holds up to IPA, bleach, and phenol wipe downs. Available irradiated, capped or click versions, and with black, blue or red ink.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorTip Style
MICPEN-01IRBlackCapped, 0.7 mm Point Tip
MICPEN-02IRBlueCapped, 0.7 mm Point Tip
Catalog number MICPEN-01IR
Price (MYR)
2,496.00
CS
Add to cart
Color:
Black
Tip Style:
Capped, 0.7 mm Point Tip
Price (MYR)
2,496.00
CS
Add to cart
Low-sodium ink
The Micronova Irradiated Cleanroom Ballpoint Pens, with low sodium ink and low particulate barrels, are designed for documentation and labeling in cleanrooms and aseptic environments. Gamma irradiated pens are suitable for Aseptic ISO 5 and other clean production areas.
  • Ball point cleanroom pens with indelible, low sodium ink
  • Designed for documentation and labeling in cleanrooms
  • White, high-density polypropylene barrel
  • Holds up to IPA, bleach and phenol wipe downs
  • Capped or Retractable options available
  • Black, blue or red ink
  • Available gamma irradiated for aseptic environments
Specifications
MaterialHDPE
RuledNo
ColorBlack
Cover ColorWhite
Cover MaterialHDPE
DescriptionCapped
Dimensions (L x W x H)5.75 x 0.375 x 0.375 in.(14.6 x 0.953 x 0.953 cm)
Length (English)5.75 in.
Length (Metric)14.6 cm
Width (English)0.375 in.
Width (Metric)0.953 cm
AutoclavableNot Autoclavable
Certifications/ComplianceISO 9001 Certified Facility; Certificate of Irradiation and Certificate of Conformance
Diameter (English) Outer0.375 in.
Diameter (Metric) Outer0.953 cm
DisposableReusable
For Use With (Application)Cleanroom and Aseptic Environment Documentation and Labeling
Packaging10 pens/bag, double bagged
Product TypeCleanroom Pen
Quantity10/Pk. 100/Cs
Size8 x 8 x 8 in.
SterilitySterile
Tip StyleCapped, 0.7 mm Point Tip
Weight (English)2 lb.
Weight (Metric)0.907 kg
Unit SizeCS