SlideMop™ Flat Head Frame and Mop Covers
SlideMop™ Flat Head Frame and Mop Covers
Actual product may vary
SlideMop™ Flat Head Frame and Mop Covers
A simple, effective mop system featuring sturdy, collapsible plates that easily slide into the quilted polyester mop cover and lock into place with an effortless downward push. Available in a choice of stainless steel or polypropylene frame.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialAutoclavable
MICMZCR63-16Polyester with Polyester FillAutoclavable
MICQSM-16PolypropyleneAutoclavable
Catalog number MICMZCR63-16
Price (MYR)
1,594.00
Add to cart
Material:
Polyester with Polyester Fill
Autoclavable:
Autoclavable
Price (MYR)
1,594.00
Add to cart
The Micronova SlideMop™ Flat Head Mop Frame and Covers is designed for use in the traditional “dip and wring” method or as a “saturation” method where liquid disposal and/or space saving is a factor.
  • Collapsible flat head mop frame and polyester covers
  • Lightweight, economical design for use throughout the facility
  • Quilted polyester mop covers with polyester pad interiors for disinfectant dispersal
  • Choice of Stainless Steel or Plastic Frame
  • Swivel joint ensures correct lift and pull or figure 8 cleaning techniques
  • For use with standard plastic or aluminum threaded handles
  • Can be used with SSU Series Stainless Steel Handle with an ACME adapter
  • Use with W-94 or W-92 wringer
  • Manufactured for use in Controlled Environments, ISO 5 thru 9
  • Available gamma irradiated: 25 kGy minimum dose
  • Good chemical resistance
  • Autoclavable
Specifications
Product TypeSlideMop, NovaPoly, Smooth Polyester, Flat Head Slip-on Mop Cover
ColorWhite
Length (English)16 in.
SterilityNon-sterile
Length (Metric)41 cm
AutoclavableAutoclavable
DisposableReusable
Care InstructionsItem may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
For Use With (Equipment)Buckets and Wringer
For Use With (Application)Cleaning and Sanitization of Cleanroom Surfaces
MaterialPolyester with Polyester Fill
Unit Size