Surface Treated Tissue Culture Dishes
Surface Treated Tissue Culture Dishes
Actual product may vary
Surface Treated Tissue Culture Dishes
Surface Treated Tissue Culture Dishes
Fisherbrand
Surface Treated Tissue Culture Dishes
Containers for cell growth, aids in sample preparation, pretreatment, storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Culture Area
LSVFB01292035 mm8.5 sq. cm
FB012925150 mm143 sq. cm
LSVFB012925150 mm143 sq. cm
FB01292160 mm21.2 sq. cm
LSVFB01292160 mm21.2 sq. cm
FB012924100 mm60.8 sq. cm
LSVFB012924100 mm60.8 sq. cm
Catalog number LSVFB012920
Price (MYR)
851.00
Add to cart
Diameter (Metric):
35 mm
Culture Area:
8.5 sq. cm
Price (MYR)
851.00
Add to cart
  • Flat bottom uniform wall thickness to ensure distortion free bottom
    • 100% virgin polystyrene, manufactured in a Class 100,000 cleanroom
  • Smooth, free surface
  • Less striation to maximize usable area for growth
  • Rim on upper side of lid mates with dish brim for easy, secure stack
  • Lids with chimbs to shape vents
  • Sterilized by gamma irradiation to SAL of 10-6
  • Non-pyrogenic
Specifications
Certifications/ComplianceCE
Closure IncludedClosure Included
Closure MaterialPolystyrene
DescriptionCell and tissue culture dishes
Bottom ShapeFlat
Culture Area8.5 sq. cm
MaterialPolystyrene
ShapeRound
SterilitySterile
Surface TreatmentStandard
TypeCell Culture Dish
For Use With (Application)Tissue culture, storage
IncludesLid
No. per Case960 Plates
No. per Pack10 Plates
Dimensions (L x W)12.5 x 37.8 mm
Diameter (Metric)35 mm
Unit Size