Heated Product Shelf Chambers
Heated Product Shelf Chambers
Actual product may vary
Heated Product Shelf Chambers
Allows drying of bulk samples, or drying of serum bottles, vials or microplates. Labconco Heated Product Shelf Chambers are wireless and removable so that they may be placed in freezer for pre-freezing.
Have Questions?
Catalog NumberIncludesVoltageHeight (Metric)Plug Type
LBC7808010503 shelves and clear chamber230 V43.1 cmBritish (UK)
Catalog number LBC780801050
Price (MYR)
62,841.00
EA
Add to cart
Includes:
3 shelves and clear chamber
Voltage:
230 V
Height (Metric):
43.1 cm
Plug Type:
British (UK)
Price (MYR)
62,841.00
EA
Add to cart
  • Short Chambers include three 10 in. diameter shelves and three temperature probes; Tall Chambers include five 10 in. diameter shelves and five temperature probes
  • Shelves are wireless and removable so that they may be placed in a freezer for pre-freezing
  • Each shelf is height adjustable to accommodate broad variety of sample sizes
  • Chamber connects to FreeZone Freeze Dryer for control and recording of shelf temperature from Lyo-Works display
  • Temperature may be set from −20° C to 60° C (−4° F to 140° F)
  • Each Chamber includes 12 in. diameter clear acrylic cylinder, vacuum release valve and 8 in. diameter opening to collector on bottom of chamber
  • Will fit 10.12 in. or 14.0 in. diameter condensers
  • For use with any FreeZone Freeze Dryer except Legacy models
Specifications
TypeHeated Shelf
No. of Shelves3
Temperature Range (Metric)-20°C to +60°C
MaterialSteel (Shelves), Acrylic (Chamber)
Certifications/ComplianceUL, CAN/CSA, CE
Includes3 shelves and clear chamber
Depth (English)15.2 in.
Depth (Metric)38.6 cm
Width (English)15.2 in.
Width (Metric)38.6 cm
Height (English)17 in.
Height (Metric)43.1 cm
Amperage2A
Plug TypeBritish (UK)
Voltage230 V
Frequency50/60 Hz
Shipping Weight (English)80 lb.
Shipping Weight (Metric)36.29 kg
Unit SizeEA