Rotary Vane Vacuum Pump
Rotary Vane Vacuum Pump
Actual product may vary
Rotary Vane Vacuum Pump
Rotary Vane Vacuum Pump designed for use with Labconco Freeze-dry Systems—compact size easily fits into freeze-dry system cabinets.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFlow RateElectrical RequirementsAmperagePlug Type
LBC7739403195 L/min.230 V, 50/60 Hz2.4 AUniversal
LBC7739402117 L/min.230 V, 50/60 Hz2.4 AReverse IEC
Catalog number LBC7739403
Price (MYR)
31,006.00
Add to cart
Flow Rate:
195 L/min.
Electrical Requirements:
230 V, 50/60 Hz
Amperage:
2.4 A
Plug Type:
Universal
Price (MYR)
31,006.00
Add to cart
Designed for use with Labconco Freeze-dry Systems—compact size easily fits into freeze-dry system cabinets
Provide a reliable vacuum source for other products including glove boxes and centrifugal concentrators.
  • Two-stage, oil-sealed pumps with sliding vane
  • Switchable high-vacuum and high-throughput modes
  • Three-position gas ballast (Closed, Low Flow, and High Flow)
  • Enclosed, fan-cooled, single-phase electric motor drives pump
  • Thermal overload protection with automatic restart
  • Inlet valve prevents air and oil suckback in the event of power or oil loss
  • Cast-aluminum housing
  • Mounted on a base plate with rubber feet for stability
Cat. No. 16-108-218 has a reverse IEC plug; models with 117L/min. pumping capacity include a retractable lifting handle; models with 195L/min. pumping capacity have a lifting bracket. A pump exhaust filter with oil and odor filter elements is also included.
Specifications
Amperage2.4 A
IncludesPump, 4 x 1 L Bottles Of Vacuum Pump Oil, 1 x Exhaust Filter with Oil Mist and Odor Filter Elements, ON/OFF Switch, Power Cord with Plug, 1 x 12.7 mm (1/2 in.) O.D. and 1 x 19.1 mm (3/4 in.) O.D. Inlet Adapter
Electrical Requirements230 V, 50/60 Hz
Dimensions (L x W x H)18.4 x 6.5 x 10.4.1 in.
MaterialCast Aluminum
Plug TypeUniversal
Flow Rate195 L/min.
For Use WithFreeze Dry Systems
Frequency50/60 Hz
Overall Height (English)10.4 in.
Overall Height (Metric)26.4 cm
Overall Width (English)6.5 in.
Pump TypeRotary Vane Vacuum Pump