Serum Bottles
Serum Bottles
Actual product may vary
Serum Bottles
Perfect for long term storage of freeze dried samples. Labconco Serum Bottles are specifically designed for lyophilization applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
LBC757321010 mL
LBC757341020 mL
Catalog number LBC7573210
Price (MYR)
2,188.00
Add to cart
Capacity (Metric):
10 mL
Price (MYR)
2,188.00
Add to cart
  • Uniform thin wall construction ensures even freezing and drying
  • Ideal containers for use in the FreeZone Stoppering Tray Dryer
  • May be connected to valve ports on drying chambers and manifolds
  • Supplied in packages of 100
Specifications
Capacity (Metric)10 mL
ColorClear
MaterialGlass
No. per Pack100/PK
SterilityNon-sterile
Product TypeSerum Bottle
Closure Size20 mm
Unit Size