Fast-Freeze™ Flask Adapter
Fast-Freeze™ Flask Adapter
Actual product may vary
Labconco
Fast-Freeze™ Flask Adapter
Fast-Freeze™ Flask Adapter for connecting Fast-Freeze™ flasks to freeze-dryer valves.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialStyleDiameter (English)
LBC7545800Borosilicate Glass45° bend0.75 in.
LBC7547200Stainless SteelStraight0.75 in.
LBC7547600Stainless Steel45° bend0.75 in.
Catalog number LBC7545800
Price (MYR)
225.00
Add to cart
Material:
Borosilicate Glass
Style:
45° bend
Diameter (English):
0.75 in.
Price (MYR)
225.00
Add to cart
For connecting Fast-Freeze™ flasks to freeze-dryer valves
  • 40 and 80 mL flasks take size 0.5 in. or 0.5 to 0.75 in.
  • Larger flasks fit size 0.75 in. or large end of 0.5 to 0.75 in.
Specifications
TypeFlask Adapter
Capacity (Metric)40, 80 mL
MaterialBorosilicate Glass
Style45° bend
For Use With (Application)For connecting fast freeze™ flasks to freeze-dryer valves
For Use With (Equipment)Fast freeze™ flask
Diameter (English)0.75 in.
Diameter (Metric)1.9 cm
Height (English)0.75 in.
Height (Metric)1.905 cm
Length (English)4.0 in.
Length (Metric)10.16 cm
Weight (English)0.5 lb.
Weight (Metric)0.2 kg
Unit Size