Rotors for CentriVap™ Systems
Rotors for CentriVap™ Systems
Actual product may vary
Rotors for CentriVap™ Systems
For use with Labconco CentriVap Systems
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsWeight (English)Weight (Metric)
LBC745500044 x 12 to 17 mL and 24 x 16 to 17 mL Tubes4 lb.1.8 kg
LBC7450700132 x 1.5 to 2 mL Microcentrifuge Tube2 lb.0.9 kg
LBC745520032 x 40 mL to 50 mL Microcentrifuge Tubes3 lb.1.4 kg
Catalog number LBC7455000
Price (MYR)
11,504.00
Add to cart
Holds:
44 x 12 to 17 mL and 24 x 16 to 17 mL Tubes
Weight (English):
4 lb.
Weight (Metric):
1.8 kg
Price (MYR)
11,504.00
Add to cart
For use with Labconco CentriVap Systems
  • Anodized aluminum rotors hold tubes at angle to maximize surface area
  • Two hole sizes on rotors 16-317-54 and 16-317-62 to -64 accommodate tubes with different O.D.
  • Maximum tube capacity for a given O.D. may vary with overall tube height
  • CentriVap DNA Systems and Proteomic come with DNA Rotor (16-317-54); Acid Systems come with Acid-Resistant 12-13 mm Rotor (16-108-450)
To prevent loss of sample during centrifugation, fill tubes only 0.33 to 0.5 full.
Specifications
MaterialAnodized Aluminum
TypeMicrocentrifuge Rotor
For Use With (Equipment)CentriVap™ DNA system
Holds44 x 12 to 17 mL and 24 x 16 to 17 mL Tubes
Volume (Metric)12 to 17 mL
Weight (English)4 lb.
Weight (Metric)1.8 kg
Unit Size