Check Order Status
Clear Drying Chamber for FreeZone™ Freeze Dryers
Actual product may vary
Clear Drying Chamber for FreeZone™ Freeze Dryers
Clear chamber provides visibility for bulk freeze drying.
Have Questions?
Change view
Catalog Number
Size
Height (English)
LBC7318702
Short
16.8 in.
LBC7318802
Short
15.3 in.
LBC7318700
Tall
24.6 in.
3 Options
Catalog number
LBC7318702
Price (MYR)
19,747.00
Add to cart
Size:
Short
Height (English):
16.8 in.
Price (MYR)
19,747.00
Add to cart
Clear chamber provides visibility for bulk freeze drying.
12.0 in. dia. acrylic chamber fits all FreeZone™ Freeze Dryers
Provides visibility for bulk freeze drying
Includes clear acrylic lid with gasket, vacuum release valve and pass-through for electrical cords
Will fit 10.12 in. or 14.0 in. diameter condensers
8 in. diameter opening to collector on bottom of chamber
Specifications
Type
Drying Chamber
No. of Shelves
3
Color
Clear
Material
Acrylic
Size
Short
For Use With (Equipment)
FreeZone™ Freeze Dryers
Diameter (English)
13.6 in.
Height (English)
16.8 in.
Shipping Weight (English)
17 lb.
Weight (English)
14 lb.
Unit Size