FlaskScrubber™ Vantage™ Glassware Washers
FlaskScrubber™ Vantage™ Glassware Washers
Actual product may vary
FlaskScrubber™ Vantage™ Glassware Washers
FlaskScrubber™ Vantage™ Glassware Washers
FlaskScrubber™ Vantage™ Glassware Washers
FlaskScrubber™ Vantage™ Glassware Washers
Versatile and efficient glassware washers designed for efficient and consistent washing of narrow-neck glassware and general labware in critical washing environments.
Have Questions?
Catalog NumberAmperage
LBC42210101316.5 A
Catalog number LBC422101013
Price (MYR)
158,452.00
EA
Add to cart
Amperage:
16.5 A
Price (MYR)
158,452.00
EA
Add to cart
Labconco FlaskScrubber™ Vantage Glassware Washers are designed for efficient and consistent washing of narrow-neck glassware and general purpose labware in critical environments such as pharmaceutical laboratories. The CleanWorks OS software is accessible through a full-color touch screen interface and provides intuitive and clear monitoring of wash loads – ensuring repeatable, effective and traceable cleaning.  Vantage models reach a maximum of 93° C wash temperature and monitor water cleanliness with a standard conductivity sensor.
FlaskScrubber Vantage models utilize HEPA filtered forced air spindle injection and SpeedVent heat-management technology to quickly dry labware. All Vantage models include a 36-place lower spindle rack, dispensing pumps for liquid detergent and acid neutralizing rinse, and a batch viewing window. Vantage models may be outfitted with a wide variety inserts and additional racks to accommodate all types of labware.
  • Touch screen display with intuitive CleanWorks OS software
  • Flexible design allows for use of Spindle Injection and Standard racks
  • Max wash temperature 93° C removes tough contaminants
  • Conductivity sensor and HEPA-filtered forced air spindle drying with SpeedVent technology
  • Durable Type 304 stainless steel chamber and racks and detergent storage cabinet
  • 36-place Spindle Injection Rack included
  • Liquid detergent and acid neutralizing rinse aid pumps included
Specifications
DescriptionFlaskScrubber Vantage Glassware Washer
DisplayFull-color Touch Screen
MaterialStainless Steel (304)
Product TypeLaboratory Glassware Washer
Flow RateHot water inlet 1.25 GPM (4.7 LPM), Cold water inlet of 3.5 GPM (13.2 LPM)
Max. Pressure120 psi (inlet)
Min. Purified Water PressureN/A, can be supplied from pressurized and non-pressurized sources
Volume (English)6.73 ft.3 chamber volume
Volume (Metric)190.6 L chamber volume
Wash Cycles12 programs with 1 to 9 total cycles
Water ConsumptionAs low as 12.9 L
StyleFreestanding FlaskScrubber Vantage Glassware Washer with Window
Certifications/ComplianceUL61010, CAN/CSA C22.2, CE
For Use With (Application)Ideal for College and University, Government, Pharmaceutical and Industrial Labs or Any Facility Utilizing Reusable Glassware
IncludesLower spindle rack
Inlet Water Temperature (English) Min.120°F
Inlet Water Temperature (English) Recommended120°F
Inlet Water Temperature (English) Max.160°F
Inlet Water Temperature (Metric) Max.71°C
Inlet Water Temperature (Metric) Min.49°C
Inlet Water Temperature (Metric) Recommended49°C
Depth (English)27.8 in.
Depth (Metric)70.6 cm
Dimensions (D x W x H) Interior27.8 x 32.2 x 32.2 to 36.1 in.
Height (English)32.2 to 36.1 in.
Height (Metric)81.8 to 91.7 cm
Width (English)32.2 in.
Width (Metric)81.8 cm
Weight (English) Shipping294 lb.
Weight (Metric) Shipping133.4 kg
Amperage16.5 A
Electrical Requirements230 V, 50/60 Hz, 16.5 A, 3 P
Voltage208/230 V