Check Order Status
Filter Disc, Surfaces Plane, Ground Cylindrical, 200 mm
Filter Disc, Surfaces Plane, Ground Cylindrical, 200 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
Have Questions?
Change view
Catalog Number
Porosity
KLC62000
0
KLC62001
1
2 Options
Catalog number
KLC62000
Price (MYR)
565.00
BX
Add to cart
Porosity:
0
Price (MYR)
565.00
BX
Add to cart
Highly resistant to most chemicals and temperatures up to 500°C
The biplane version may be required in specific installation circumstances
Specifications
Material
Borosilicate Glass (3.3)
Sterility
Non-sterile
No. per Pack
1 Ea.
Porosity
0
Thickness (Metric)
11.5 mm ± 0.5 mm
Diameter (Metric)
200 mm
Unit Size
BX