Kimtech™ Pure G3 White Nitrile Gloves
Kimtech™ Pure G3 White Nitrile Gloves
Actual product may vary
Kimtech™ Pure G3 White Nitrile Gloves
Hand-specific Kimtech™ Pure G3 white nitrile gloves.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC568907
KLC56888X-Small, 6
KLC568938.5
Catalog number KLC56890
Price (MYR)
1,478.00
Add to cart
Size:
7
Price (MYR)
1,478.00
Add to cart
200 pairs of Kimtech™ Pure G3 white nitrile gloves. Hand-specific gloves.
  • Provides enhanced sterile cleanroom protection against a wide range of contaminants, chemicals, viruses, and cytotoxic drugs.
  • With an AQL of 1.5, the anti-static tested, latex and silicon free gloves feature a textured finish for improved grip and sensitivity while being suitable for double donning.
  • Validated to a Sterility Assurance Level (SAL) of 10-6, performing with a cleanliness level of a maximum of 950 particles >0.5μm/cm2, contains low endotoxins, meets FDA food contact and USP 800 guidelines.
  • The hand-specific gloves are cleanroom-ready; double-packaged in polyethylene bags to ensure your processes and workflows can continue operating efficiently within ISO 3 level cleanrooms and higher including Biotech, Aseptic processing, Parenteral drug and Ophthalmic manufacturing environments.
Specifications
Certifications/ComplianceASTM D6978-05
AQL1.5
ColorWhite
DisposableSingle-use
Exterior FinishFully Textured
HandSpecific
Latex FreeYes
MaterialNitrile
PowderedPowder Free
Size7
SterilitySterile
Thickness (Metric)5.1 mm
Length (English)12 in.
Unit Size