Kimtech™ Purple Nitrile-Xtra™ Gloves
Kimtech™ Purple Nitrile-Xtra™ Gloves
Actual product may vary
Kimtech™ Purple Nitrile-Xtra™ Gloves
Kimtech™ Purple Nitrile-Xtra™ Gloves
Kimtech™ Purple Nitrile-Xtra™ Gloves
Kimtech™ Purple Nitrile-Xtra™ Gloves
Purple Nitrile Xtra™ Gloves are 12 in. in length with a beaded cuff to provide maximum protection for higher-risk procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC50601Small
KLC50602Medium
KLC50603Large
Catalog number KLC50601
Price (MYR)
662.00
Add to cart
Size:
Small
Price (MYR)
662.00
Add to cart
Purple Nitrile Xtra™ Gloves are 12 in. in length with a beaded cuff to provide maximum protection for higher-risk procedures.
  • Textured fingertips offers dexterity and tactile sensitivity
  • Latex-free
  • Ambidextrous
  • 12 in. length
  • Beaded cuff prevent roll-down and allow for easy donning
  • Powder free
Specifications
Certifications/ComplianceASTM D6978-05 (non-sterile only), Meets USP <800> guidelines, Meets NFPA standard 1999-2018
Chemotherapeutic RatedYes
ApplicationsHealthcare, Laboratory
AQL0.65
ColorPurple
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)6 mil
Finger Thickness (Metric)0.15 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)4.7 mil
Palm Thickness (Metric)0.11 mm
PowderedPowder Free
SizeSmall
Thickness (English)6 mil
Thickness (Metric)0.15 mm
FDA 510 (k)Yes
Model50601
SeriesKimtech™
Tensile Strength21 MPa
Length (English)12 in.
Length (Metric)30.5 cm
Unit Size