Prefabricated Quick-Disconnect Safety Disposal Cans
Prefabricated Quick-Disconnect Safety Disposal Cans
Actual product may vary
Prefabricated Quick-Disconnect Safety Disposal Cans
For continuous collection of HPLC waste flow
Have Questions?
Catalog NumberCapacity (English)Diameter (Metric) OuterHeight (Metric)Includes
JUSBY127522 gal.30.5 cm37.5 cmSS/PP Fittings
Catalog number JUSBY12752
Price (MYR)
4,358.00
EA
Add to cart
Capacity (English):
2 gal.
Diameter (Metric) Outer:
30.5 cm
Height (Metric):
37.5 cm
Includes:
SS/PP Fittings
Price (MYR)
4,358.00
EA
Add to cart
For continuous collection of HPLC waste flow
Polyethylene can with quick-disconnect system permits “hands-free” liquid waste accumulation for maximum protection against chemicals. Suitable for pump-fed or gravity flow HPLC waste methods.
  • Allows free flow of liquid from outside source into FM-approved polyethylene disposal can
  • Two fittings accommodate tubing with 0.25 to 0.375 in. diameter (0.6 to 1.0cm)
  • Built-in pressure-relief prevents excessive pressure in extreme temperatures
  • Translucent white color makes detection of liquid level easy
  • Available with stainless-steel fitting with PTFE O-rings or economical polypropylene fitting with EPDM O-rings
Specifications
Capacity (Metric)8 L
Certifications/ComplianceFM, OSHA, EPA
Diameter (Metric) Outer30.5 cm
Diameter (English) Outer12 in.
Capacity (English)2 gal.
ColorWhite
Height (Metric)37.5 cm
Quantity1 Ea.
Diameter (English) Opening3.65 in.
Diameter (Metric) Opening9.3 cm
IncludesSS/PP Fittings
For Use With (Application)For continuous collection of HPLC waste flow
Height (English)14.75 in.
MaterialPolyethylene
Product TypeSafety Disposal Can
Unit SizeEA