CleanTip™ Swabs: Absorbond™ Series
CleanTip™ Swabs: Absorbond™ Series
Actual product may vary
CleanTip™ Swabs: Absorbond™ Series
No contaminating adhesives
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConstructionDiameter (Metric) TipProduct TypeLength (Metric) Handle
ITWTX759BFlexible head paddle, compact handle3.2mmSwab59.9mm
ITWTX762Flexible head paddle, long handle6.6mmSwab145.2mm
Catalog number ITWTX759B
Price (MYR)
1,046.88
Add to cart
Construction:
Flexible head paddle, compact handle
Diameter (Metric) Tip:
3.2mm
Product Type:
Swab
Length (Metric) Handle:
59.9mm
Price (MYR)
1,046.88
Add to cart
No contaminating adhesives
Perfect for abrasion-sensitive applications requiring very low residue such as wiping sensitive-coated optics surfaces.
  • Double-layered, hydroentangled, continuous-filament polyester heads thermally molded onto polypropylene handles without adhesives
  • Ultraclean head materials are precision molded so that every swab is constructed to exacting tolerances
  • Long filaments reduce risk of fiber release; hydroentanglement lowers nonvolatile residue to a very low level
  • Heads are heat molded onto polypropylene handles and perimeter edges heat sealed to prevent fiber release
  • Excellent chemical resistance; can be used with a wide range of solvents
  • Constructed from the cleanest materials available and manufactured in a cleanroom using state-of-the-art processes and equipment
  • High-precision automated process guarantees consistent performance, sizing and cleanliness
  • Silicone-free Bag-Within-A-Bag™ packaging consists of a solvent-resistant, thermally sealed exterior bag that protects the inner bags containing the swabs
  • Each outer bag is individually lot-coded for easy traceability and quality control
Specifications
ConstructionFlexible head paddle, compact handle
Diameter (English) Handle0.08 in.
Length (English)2.36 in.
For Use With (Application)Cleaning sensitive surfaces such as optical assemblies, solvent use (such as IPA), cleaning of grooves, tracks, slots and other small spaces
Handle ColorLight Green
Product TypeSwab
Diameter (Metric) Handle2mm
Length (Metric)60mm
Length (English) Handle2.36 in.
Length (Metric) Handle59.9mm
Thickness (Metric)1.4mm
Length (English) Tip0.39 in.
Length (Metric) Tip9.9mm
Width (Metric) Tip1.97cm
Width (English) Tip0.13 in.
Diameter (Metric) Tip3.2mm
Diameter (English) Tip0.12 in.
Adhesive TypeThermal
Handle MaterialPolypropylene
Tip MaterialHydroentangled Absorbond™ Polyester
Unit Size