Alpha™ Polyester Series Swabs
Alpha™ Polyester Series Swabs
Actual product may vary
Alpha™ Polyester Series Swabs
Knitted and cleanroom laundered polyester head provides a neutral background for obtained sample because of its high cleanliness levels. Texwipe™ Alpha™ Swabs are widely recommended for surface sampling and cleaning validation work.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConstructionThickness (Metric)DescriptionLength (Metric)
ITWTX754BRigid head core, compact handle3.0mmMini Alpha™ Swab69.5mm
ITWTX761Flexible head paddle, long handle2.8mmAlpha™ Swab with Long Handle162.3mm
Catalog number ITWTX754B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Construction:
Rigid head core, compact handle
Thickness (Metric):
3.0mm
Description:
Mini Alpha™ Swab
Length (Metric):
69.5mm
Knitted and cleanroom laundered polyester head provides a neutral background for obtained sample because of its high cleanliness levels. Texwipe™ Alpha™ Swabs are widely recommended for surface sampling and cleaning validation work.
Two layers of cleanroom-laundered AlphaLite™ polyester tips thermally sealed onto polypropylene handles.
  • Ultraclean head materials are precision molded so that every swab is constructed to exacting tolerances
  • No contaminating adhesives
  • Superior resistance to wide range of chemicals and solvents
  • Ultralow releasable particulates, ions, and NVR
  • Recommended for cleaning validation
  • Excellent sorbency and release properties
  • Broad range of tip and handle configurations
  • Constructed from the cleanest materials available and manufactured in a cleanroom using state-of-the-art processes and equipment
  • High-precision automated process guarantees consistent performance, sizing and cleanliness Silicone-free Bag-Within-A-Bag™ packaging consists of a solvent-resistant, thermally sealed exterior bag that protects the inner bags containing the swabs Each outer bag is individually lot-coded for easy traceability and quality control
Specifications
DescriptionMini Alpha™ Swab
ConstructionRigid head core, compact handle
For Use With (Application)Precision spot cleaning of grooves, tracks, slots and other small spaces, applying and removing lubricants, adhesives and other solutions in a critical clean environment, solvent use (such as IPA)
Handle ColorLight Green
Diameter (Metric) Handle2.5mm
Length (Metric)69.5mm
Length (Metric) Handle60mm
MaterialPolyester
Thickness (Metric)3.0mm
Length (Metric) Tip9.5mm
Width (Metric) Tip3.5mm
Diameter (Metric) Tip3.5mm
Adhesive TypeThermal
Handle MaterialPolypropylene
Tip MaterialKnitted Alpha™ Polyester
Unit Size