CleanTip™ Swabs: CleanFoam™ Series
CleanTip™ Swabs: CleanFoam™ Series
Actual product may vary
CleanTip™ Swabs: CleanFoam™ Series
Excellent sorbency
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConstructionLength (English)SizeThickness (Metric)
ITWTX740BFlexible head paddle, long handle6.41 in.Medium5mm
FIS19-034-190Precision pointed tip, compact handle2.63 in.Small3.5mm
Catalog number ITWTX740B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Construction:
Flexible head paddle, long handle
Length (English):
6.41 in.
Size:
Medium
Thickness (Metric):
5mm
Excellent sorbency
Laundered CleanFoam heads constructed of 100 pores per inch open-cell polyurethane foam for excellent capacity and cushioning.
  • Cleanest foam swabs available with superior solvent capacity and contamination capture
  • Solvent-processed for very low levels of nonvolatile residue and particle generation
  • Good chemical resistance—can be used with solvents such as isopropyl alcohol and applying lubricants and other liquids
  • Heads thermally welded onto polypropylene handles, without any contaminating adhesivesConstructed from the cleanest materials available
  • High-precision automated process guarantees consistent performance, sizing and cleanliness
  • Silicone-free Bag-Within-A-Bag™ packaging consists of a solvent-resistant, thermally sealed exterior bag that protects the inner bags containing the swabs
  • Each outer bag is individually lot-coded for easy traceability and quality control
Specifications
ConstructionFlexible head paddle, long handle
Diameter (English) Handle0.126 in.
Length (English)6.41 in.
For Use With (Application)Applying and removing lubricants, adhesives and other solutions in a critical clean environment, scrubbing recessed areas, removal of excess materials, debris, cleaning intersecting surfaces and joints
Handle ColorLight Green
Product TypeSwab
Diameter (Metric) Handle3.2mm
Length (Metric)163mm
Length (English) Handle5.74 in.
Length (Metric) Handle146mm
MaterialCleanFoam
SizeMedium
Thickness (Metric)5mm
Length (English) Tip0.669 in.
Length (Metric) Tip17mm
Width (Metric) Tip2.3mm
Width (English) Tip0.134 in.
Diameter (Metric) Tip6.4mm
Diameter (English) Tip0.252 in.
Adhesive TypeThermal
Handle MaterialPolypropylene
Tip StyleFlexible
Tip Material100 ppi CleanFoam™ Polyurethane Foam
Unit Size