Vertex™ Sterile High Sorption Wipers
Vertex™ Sterile High Sorption Wipers
Actual product may vary
Vertex™ Sterile High Sorption Wipers
100% continuous-filament, polyester knit fabric wipers made in the USA from virgin stock
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)Description
ITWTX30499 in.Dry
ITWTX3049P9 in.Pre-wetted
Catalog number ITWTX3049
Price (MYR)
1,026.77
Add to cart
Length (English):
9 in.
Description:
Dry
Price (MYR)
1,026.77
Add to cart
Sealed-edge sterile wipers, dry or pre-wetted
  • Soft feel with high sorption capacity
  • Confirm radiation dose met specified dose at time of receiving
  • Processed on Texwipe's Vertex™ manufacturing system
  • Quarterly microbial testing by Agarose Overlay and Limulus Amebocyte Lysate (LAL) endotoxin testing
Pre-Wetted Wipers
  • 70% USP-Grade IPA/30% USP Purified Water
Specifications
Product TypeHigh Sorption Wiper
Length (English)9 in.
Length (Metric)22.9 cm
Width (English)9 in.
Width (Metric)22.9 cm
MaterialPolyester
Certifications/ComplianceISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
DescriptionDry
EdgesSealed
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)Ideal for cleaning delicate surfaces or applications