PetriPile
PetriPile
Actual product may vary
PetriPile
Stainless steel storage rack for Petri dishes.
Have Questions?
Catalog NumberDiameter (Metric) Inner
ISC24105555 mm
Catalog number ISC241055
Price (MYR)
1,063.00
BX
Add to cart
Diameter (Metric) Inner:
55 mm
Price (MYR)
1,063.00
BX
Add to cart
PetriPile ensures safe handling, carrying and storage of Petri dishes. Its ergonomics make the loading of your incubator easier and gives overall visibility of your Petri dishes.
  • Capacity: 36 Petri dishes
  • Autoclavable and stackable
  • All stainless steel
Available for Petri dishes with the following diameters:
  • 55 mm
  • 65 mm
  • 90 mm
  • 150 mm
Specifications
Product TypePetri Dish Holder
AutoclavableAutoclavable
ColorSilver
HoldsPetri Dishes
MaterialStainless Steel
ShapeRectangular
StackableYes
SterilityNon-sterile
Width (Metric)21.5 cm
Width (English)8.4 in.
Height (Metric)13 cm
Height (English)5.1 in.
Diameter (Metric) Inner55 mm
Unit SizeBX