MPC 104 Tp Vacuum Pump
MPC 104 Tp Vacuum Pump
Actual product may vary
MPC 104 Tp Vacuum Pump
For non-volatile solvents and acids
Have Questions?
Catalog NumberFlow Rate
ILM412443-0215 L/min.
Catalog number ILM412443-02
Price (MYR)
15,277.00
EA
Add to cart
Flow Rate:
15 L/min.
Price (MYR)
15,277.00
EA
Add to cart
Tp flow rate 15L/min for non-volatile solvents and acids 230V 50/60Hz 235mm(w) x 140mm(d) x 340mm(h) vacuum >2mbar Ilmvac
Specifications
Electrical Requirements230 V 50/60 Hz
Depth (Metric)140 mm
ModelMPC 104 T
Weight (Metric)7.1 kg
Flow Rate15 L/min.
Frequency50/60 Hz
Overall Height (Metric)340 mm
Overall Width (Metric)235 mm
Pump TypeVacuum Pump
Maximum Pressure<2 mbar