Sterilin™ Plastic Round Base Tubes
Sterilin™ Plastic Round Base Tubes
Actual product may vary
Sterilin™ Plastic Round Base Tubes
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene.
Have Questions?
Catalog NumberTubing SizePackaging Quantity
IKART150.6 mL1000
Catalog number IKART15
Price (MYR)
45,048.00
EA
Add to cart
Tubing Size:
0.6 mL
Packaging Quantity:
1000
Price (MYR)
45,048.00
EA
Add to cart
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene and supplied nonsterile.
Not available in all geographic regions.
  • Suitable for samples ranging from 0.6 to 11mL
  • Supplied nonsterile
Specifications
DescriptionRound Base Plastic Tubes
MaterialPolystyrene
Diameter (Metric) Outer6 mm
Packaging Quantity1000
Product LineSterilin™
SterilityNon-sterile
Tubing Size0.6 mL
Product TypeBase Tube
Unit SizeEA