Check Order Status
Model AS 130.1 Universal Attachment
Actual product may vary
Model AS 130.1 Universal Attachment
Have Questions?
Catalog number
IKA8017300
Price (MYR)
3,719.00
EA
Add to cart
Price (MYR)
3,719.00
EA
Add to cart
Specifications
Product Type
Universal Attachment
Dimensions (L x W x H)
234 x 325 x 88 mm
For Use With (Equipment)
KS130 Shaker