Fixed Needle Autosampler Syringes, 2500uL, for Thermo Scientific™ Instruments
Fixed Needle Autosampler Syringes, 2500uL, for Thermo Scientific™ Instruments
Actual product may vary
Fixed Needle Autosampler Syringes, 2500uL, for Thermo Scientific™ Instruments
Gas-tight, headspace.
Have Questions?
Catalog NumberVolume (Metric)
HYP365Q21312.5 mL
Catalog number HYP365Q2131
Price (MYR)
3,129.00
EA
Add to cart
Volume (Metric):
2.5 mL
Price (MYR)
3,129.00
EA
Add to cart
Gas-tight, headspace.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description2.5 mL FN GT HS, 23 G, 65 mm, 115°C
Needle Gauge23 G
Length (Metric)65 mm
Product TypeGC Autosampler Syringe
Max. Temperature (Metric)115°C
QuantityEa.
Volume (Metric)2.5 mL
Length (Metric) Needle65 mm
Needle Point StyleSide Hole
For Use With (Equipment)TriPlus RSH Autosampler
PTFE Tipped PlungerYes
Unit SizeEA