Cimarec+™ Hotplate Series
Cimarec+™ Hotplate Series
Actual product may vary
Thermo Fisher Scientific
Cimarec+™ Hotplate Series
Get exceptional safety and temperature performance for your routine protocols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeTop Plate MaterialPlug Type
HP888541054.25 x 4.25 in.CeramicEU-Style, UK-Style, AUS-Style, CN-Style
HP888571057.25 x 7.25 in.CeramicEU-Style, UK-Style, AUS-Style, CN-Style
HP888571087.25 x 7.25 in.AluminumEU-Style, UK-Style, AUS-Style, CN-Style
HP8885010510.25 x 10.25 in.CeramicEU-Style, UK-Style, AUS-Style, CN-Style
Catalog number HP88854105
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
4.25 x 4.25 in.
Top Plate Material:
Ceramic
Plug Type:
EU-Style, UK-Style, AUS-Style, CN-Style
Get exceptional safety and temperature performance for your routine protocols with the Thermo Scientific™ Cimarec+™ Hotplate series. You'll find exceptional safety and temperature performance for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operations.
Advanced technology
  • Temperature adjustable in 1 degree increments
  • Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F)
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Warranty: 3 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Dimensions (L x W x H)4.25 in. x 4.25 in.
TypeHot Plate
Certifications/ComplianceCOA, COC
DisplayLED
Plug TypeEU-Style, UK-Style, AUS-Style, CN-Style
ShapeSquare
Voltage230 V
Warranty3 Years
Top Plate MaterialCeramic
Size4.25 x 4.25 in.
Temperature (Metric)Ambient to 540°C
Height (English)3.6 in.
Length (English)10 in.
Weight (English)6.8 lb.
Width (English)5 in.
Height (Metric)9.1 cm
Length (Metric)25.4 cm
Weight (Metric)3.1 kg
Width (Metric)12.7 cm
Electrical Requirements230 V, 50/60 Hz
Frequency50/60 Hz
Includes(3) Power Cords
Unit Size