SuperNuova+™ Hotplate Series
SuperNuova+™ Hotplate Series
Actual product may vary
SuperNuova+™ Hotplate Series
SuperNuova+™ Hotplate Series
Thermo Fisher Scientific
SuperNuova+™ Hotplate Series
Get reliable performance, dependable safety and simple operation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeTop Plate MaterialElectrical Requirements
HP8885019510.25 x 10.25 in.Ceramic230 V, 50/60 Hz
HP888571957.25 x 7.25 in.Ceramic230 V, 50/60 Hz
Catalog number HP88850195
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
10.25 x 10.25 in.
Top Plate Material:
Ceramic
Electrical Requirements:
230 V, 50/60 Hz
Our new Thermo Scientific™ SuperNuova+™ hotplate series provides reliable performance, dependable safety and simple operation. Units are available in two sizes, including a multi-position model with optimized controls and settings for applications that demand advanced precision.
Intuitive design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating
  • Raised display design protects electronics from spills
  • Ability to save temperature setting
  • Optional PT1000 temperature probe and splash guard protection shield
  • Includes: PT100 Temperature Probe, Rod and Clamps
Advanced technology
  • Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F)
Warranty: 3 year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Dimensions (L x W x H)16.2 x 11.3 x 4 in. (41.1 x 28.7 x 10.2cm)
TypeHot Plate
Certifications/CompliancecCSAus, CE
DisplayLED
Plug TypeEU-Style, UK-Style, AUS-Style, CN-Style
ShapeSquare
Voltage230 V
Warranty3 Years
Top Plate MaterialCeramic
Size10.25 x 10.25 in.
Temperature (Metric)Ambient to 400°C
Height (English)4 in.
Length (English)16.2 in.
Weight (English)16 lb.
Width (English)11.3 in.
Height (Metric)10.2 cm
Length (Metric)41.1 cm
Weight (Metric)7.3 kg
Width (Metric)28.7 cm
Electrical Requirements230 V, 50/60 Hz