Owl™ HEP Series Semidry Electroblotting Systems
Owl™ HEP Series Semidry Electroblotting Systems
Actual product may vary
Thermo Fisher Scientific
Owl™ HEP Series Semidry Electroblotting Systems
Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDimensions (L x W x H)
HEP-1For 1 mini gel: Transfer area 20 x 20cm9.86 x 9.86 x 2 in. (25 x 25 x 5 cm)
HEP-3For 4 mini gels: Transfer area 35 x 45cm19.88 x 15.63 x 2 in. (50.4 x 39.7 x 5.08 cm)
Catalog number HEP-1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
For 1 mini gel: Transfer area 20 x 20cm
Dimensions (L x W x H):
9.86 x 9.86 x 2 in. (25 x 25 x 5 cm)
Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems. Transfers can be done in as little as 15 minutes.
  • Fast, efficient transfer of proteins and nucleic acids to membranes
  • Buffer contained within blotting paper is sufficient for efficient transfer of molecules
  • Base contains stainless-steel cathode plate; lid has platinum/titanium anode plate
  • Ideal for proteins less than 150 kDa
Includes:
  • Base and lid
  • Power supply leads
  • Heavy-duty knobs
Specifications
DescriptionFor 1 mini gel: Transfer area 20 x 20cm
Gel SizeMini (20 cm x 20 cm)
Product LineOwl
TypeElectroblotting System
Dimensions (L x W x H)9.86 x 9.86 x 2 in. (25 x 25 x 5 cm)
Unit Size