Stainless Steel Bath Cover
Stainless Steel Bath Cover
Actual product may vary
Stainless Steel Bath Cover
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Bath Cover for Grant Stirred Water Baths is available flat or gabled with removable hinge.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
GRASTL12ST12 tank
GRASTL38ST38 tank
Catalog number GRASTL12
Price (MYR)
3,300.00
Add to cart
For Use With (Equipment):
ST12 tank
Price (MYR)
3,300.00
Add to cart
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Bath Cover for Grant Stirred Water Baths is available flat or gabled with removable hinge.
  • Stainless steel construction
  • Plastic handle
  • Available flat (STL 5), or gabled with removable hinge (STL 12, STL 38)
  • For use with Grant Stirred Water Baths and Heated Circulators
  • Prevents heat from escaping
  • Reduces sample contamination
Specifications
For Use With (Equipment)ST12 tank