Stainless Steel Tank and Bridge Plate for Baths
Stainless Steel Tank and Bridge Plate for Baths
Actual product may vary
Stainless Steel Tank and Bridge Plate for Baths
Combine with any Grant Heating Circulator or Bath. Grant Instruments™ Stainless Steel Tank and Bridge Plate for Baths has a temperature range of –15 to 200°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)For Use With (Equipment)
GRAST3838 LOptima range immersion thermostats - T100, TC120, TXF150 or TXF200
GRAST55 LOptima range immersion thermostats - T100, TC120, TXF150 or TXF200
Catalog number GRAST38
Price (MYR)
13,876.00
EA
Add to cart
Capacity (Metric):
38 L
For Use With (Equipment):
Optima range immersion thermostats - T100, TC120, TXF150 or TXF200
Price (MYR)
13,876.00
EA
Add to cart
Combine with any Grant Heating Circulator or Bath. Grant Instruments™ Stainless Steel Tank and Bridge Plate for Baths has a temperature range of –15 to 200°C.
  • High efficiency stainless steel construction
  • Templerature range: -15 to 200°C
  • Includes integral raised feet for ease of carrying, repositioning, and retort access
Specifications
Capacity (Metric)38 L
For Use With (Equipment)Optima range immersion thermostats - T100, TC120, TXF150 or TXF200
Temperature Range15°C to 200°C
MaterialStainless Steel
Length (Metric)730 mm
Unit SizeEA