Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA610350010
Price (MYR)
1,839.00
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
1,839.00
Add to cart
Chemical Identifiers
CAS6980-08-1
IUPAC Name4-chloro-3-nitropyridin-2-amine
Molecular FormulaC5H4ClN3O2
InChI KeyDIRINUVNYFAWQF-UHFFFAOYSA-N
SMILESNC1=NC=CC(Cl)=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%