Aluminum oxide, for chromatography, basic, Brockmann I, 50-200 μm, 90A
Aluminum oxide, for chromatography, basic, Brockmann I, 50-200 μm, 90A
Thermo Scientific Chemicals
Aluminum oxide, for chromatography, basic, Brockmann I, 50-200 μm, 90A
CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Catalog NumberQuantity
FSA4633500101 kg
Catalog number FSA463350010
Price (MYR)
817.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
817.00
EA
Add to cart
Specifications
Boiling Point2977.0°C
Chemical Name or MaterialAluminum oxide
Melting Point2030.0°C
CAS1344-28-1
Quantity1 kg