Beclomethasone dipropionate, 97%, Thermo Scientific Chemicals
Beclomethasone dipropionate, 97%, Thermo Scientific Chemicals
Beclomethasone dipropionate, 97%, Thermo Scientific Chemicals
Beclomethasone dipropionate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Beclomethasone dipropionate, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA460990010
Price (MYR)
1,607.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,607.00
Add to cart
Specifications
Chemical Name or MaterialBeclomethasone dipropionate
CAS5534-09-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May damage fertility or the unborn child.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.