Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Thermo Scientific Chemicals
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
CAS: 14552-35-3 | C4H18CuN4O2 | 217.76 g/mol
Have Questions?
Catalog NumberQuantity
FSA4606800101 L
Catalog number FSA460680010
Price (MYR)
428.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
428.00
Add to cart
Chemical Identifiers
CAS99-64-9
IUPAC Name3-(dimethylamino)benzoic acid
Molecular FormulaC9H11NO2
InChI KeyNEGFNJRAUMCZMY-UHFFFAOYSA-N
SMILESCN(C)C1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point150°C to 153°C
Titration with NaOH>=97.5 %