Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 112-39-0 | C17H34O2 | 270.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA460390010
Price (MYR)
302.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
302.00
Add to cart
Chemical Identifiers
CAS35794-11-7
IUPAC Name(3R,5S)-3,5-dimethylpiperidin-1-ium
Molecular FormulaC7H16N
InChI KeyIDWRJRPUIXRFRX-KNVOCYPGSA-O
SMILESC[C@H]1C[NH2+]C[C@@H](C)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=95 % (sum of cis and trans)
Refractive index1.4425 to 1.4475 (20°C, 589 nm)