Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 112-62-9 | C19H36O2 | 296.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA460380050
Price (MYR)
964.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
964.00
Add to cart
Chemical Identifiers
CAS109-01-3
IUPAC Name1-methylpiperazine
Molecular FormulaC5H12N2
InChI KeyPVOAHINGSUIXLS-UHFFFAOYSA-N
SMILESCN1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.4635-1.4675 @ 20?C
Appearance (Color)Clear colorless to pale yellow