Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 112-62-9 | C19H36O2 | 296.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA460380010
Price (MYR)
372.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
372.00
Add to cart
Chemical Identifiers
CAS35293-35-7
IUPAC Namechloro(2-methyl-2-phenylpropyl)magnesium
Molecular FormulaC10H13ClMg
InChI KeyBXKYFONHSSFLKR-UHFFFAOYSA-M
SMILESCC(C)(C[Mg]Cl)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearanceatmosphere
Appearance (Color)Colorless to yellow to grey
AppearanceIf precipitation is present, gently mix
AppearanceMay develop some turbidity or precipitate