Methyl myristate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl myristate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl myristate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl myristate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl myristate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 124-10-7 | C15H30O2 | 242.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA460360010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS2226-96-2
IUPAC Name(4-hydroxy-2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Molecular FormulaC9H18NO2
InChI KeyUZFMOKQJFYMBGY-UHFFFAOYSA-N
SMILESCC1(C)CC(O)CC(C)(C)N1[O]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)Orange
Infrared spectrumConforms
Melting point69°C to 73°C
GC>=97.5 %