Mifepristone, 98%, Thermo Scientific Chemicals
Mifepristone, 98%, Thermo Scientific Chemicals
Mifepristone, 98%, Thermo Scientific Chemicals
Mifepristone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mifepristone, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number FSA459980010
Price (MYR)
503.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
503.00
Add to cart
Specifications
Chemical Name or MaterialMifepristone
CAS84371-65-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May damage fertility or the unborn child.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.