Risperidone, 99%, Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
CAS: 106266-06-2 | C23H27FN4O2 | 410.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA459390010
Price (MYR)
402.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
402.00
Add to cart
Chemical Identifiers
CAS121-33-5
IUPAC Name4-hydroxy-3-methoxybenzaldehyde
Molecular FormulaC8H8O3
InChI KeyMWOOGOJBHIARFG-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMaterial sourced in the U.S. and in other countries
FormCrystals or crystalline powder
Appearance (Color)White to cream or pale yellow
Assay (GC)≥98.5% (non-U.S. sourced material)
CommentMaterial sourced in the U.S. and in other countries