βmethasone, 97%
βmethasone, 97%
βmethasone, 97%
Thermo Scientific Chemicals
βmethasone, 97%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number FSA459070010
Price (MYR)
378.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
378.00
Add to cart
Chemical Identifiers
CAS103-47-9
IUPAC Namesodium 2-(cyclohexylamino)ethane-1-sulfonate
Molecular FormulaC8H16NNaO3S
InChI KeyTXWKJOVDGFRVOG-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CCNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
UV(0.1 M in water)
Appearance (Form)Crystalline powder
Water=<1 % (K.F.)