Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
CAS: 79-03-8 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA458970250
Price (MYR)
226.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
226.00
Add to cart
Chemical Identifiers
CAS462-06-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4643 to 1.4663 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.1 % (K.F.)