Simvastatin, 98%, Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
CAS: 79902-63-9 | C25H38O5 | 418.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number FSA458840010
Price (MYR)
318.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
318.00
Add to cart
Specifications
Chemical Name or MaterialSimvastatin
CAS79902-63-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
Obtain special instructions before use.
IF exposed or concerned:
View more