Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number FSA458690010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Titration excess NaOH>=99.5 %
Iron (Fe)=<5 ppm
Appearance (Form)Crystals
Residue after ignition=<0.01 % (as SO4)