Ammonium hydroxide, Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number FSA458680010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS453-20-3
IUPAC Nameoxolan-3-ol
Molecular FormulaC4H8O2
InChI KeyXDPCNPCKDGQBAN-UHFFFAOYNA-N
SMILESOC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Appearance (Form)Liquid