Econazole, 99%, Thermo Scientific Chemicals
Econazole, 99%, Thermo Scientific Chemicals
Econazole, 99%, Thermo Scientific Chemicals
Econazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Econazole, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA458630050
Price (MYR)
302.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
302.00
Add to cart
Specifications
Chemical Name or MaterialEconazole
CAS27220-47-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful if swallowed.
Health Hazard 3GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.