Omeprazole, 98+%
Omeprazole, 98+%
Omeprazole, 98+%
Thermo Scientific Chemicals
Omeprazole, 98+%
CAS: 73590-58-6 | C17H19N3O3S | 345.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA458520010
Price (MYR)
293.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
293.00
Add to cart
Chemical Identifiers
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.05 %
Methanol=<0.1 %
Cobalt (Co)=<0.02 ppm
Iron (Fe)=<0.1 ppm